missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C6orf136 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06307PCP
This item is not returnable.
View return policy
Description
C6orf136 Polyclonal antibody specifically detects C6orf136 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| C6orf136 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| chromosome 6 open reading frame 136, Em:AB023049.8, hypothetical protein LOC221545, MGC15854 | |
| Produced in rabbits immunized with E. coli-derived Human C6orf136 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 221545 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction