missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abhd5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 26 publications
€ 226.00 - € 497.00
Specifications
| Antigen | Abhd5 |
|---|---|
| Dilution | Western Blot 1:500, Simple Western 1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18139719
|
Novus Biologicals
NB110-41576SS |
0.025 mL |
€ 226.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18778183
|
Novus Biologicals
NB110-41576 |
0.1 mL |
€ 497.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Abhd5 Polyclonal specifically detects Abhd5 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.Specifications
| Abhd5 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Lipid and Metabolism, Lipid Droplets, Signal Transduction | |
| PBS with 0.02% Sodium Azide | |
| abhydrolase domain containing 5, Abhydrolase domain-containing protein 5,1-acylglycerol-3-phosphate O-acyltransferase ABHD5, CDS, CGI58, EC 2.3.1.51, IECN2, Lipid droplet-binding protein CGI-58, MGC8731, NCIE2CGI-58 | |
| ABHD5 | |
| IgG | |
| IgG purified | |
| Human and rat. Mouse reactivity reported in scientific literature (PMID:23878361). |
| Western Blot 1:500, Simple Western 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Q8WTS1 | |
| 51099 | |
| A synthetic peptide made to an internal region within residues 200-300 of the human Abhd5 protein. [Swiss-Prot# Q8WTS1] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title