missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABHD8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5111464
This item is not returnable.
View return policy
Description
Immunogen sequence: LGYEVVAPDL AGHGASSAPQ VAAAYTFYAL AEDMRAIFKR YAKKRNVLIG HSYGVSFCTF LAHEYPDLVH KVIMINGGGP TALEPSFCSI Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.
The alpha/beta hydrolase superfamily comprise diverse members that are involved in important biochemical processes and related to various diseases. They have unrelated sequences, various substrates, and different kinds of catalytic activities, yet they share the same canonical alpha/beta hydrolase fold, which consists of an eight-stranded parallel alpha/beta structure. They are also characterized by a catalytic triad composed of a histidine, an acid and a nucleophile. Members of this superfamily are often drug targets for treating diseases, such as diabetes, Alzheimer's disease, obesity and blood clotting disorders. The alpha/beta hydrolase domain containing (ABHD) gene subfamily is comprised of 15 mostly uncharacterized members, most of which utilize a serine nucleophile to form the G-X-S-X-G nucleophile elbow. ABHD8 is a 439 amino acid protein that belongs to the AB hydrolase superfamily.
Specifications
| ABHD8 | |
| Polyclonal | |
| Unconjugated | |
| ABHD8 | |
| 0910001L24Rik; AB030191; ABHD8; abhydrolase domain containing 8; abhydrolase domain-containing protein 8; Alpha/beta hydrolase domain-containing protein 8; protein ABHD8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79575 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96I13 | |
| ABHD8 | |
| Recombinant Protein corresonding to Human ABHD8. Recombinant protein control fragment (Product #RP-105097). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction