missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ABHD9 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16647874
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16647874 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16647874 Supplier Invitrogen™ Supplier No. PA5101397

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ABHD9.

The alpha/beta hydrolase superfamily comprise diverse members that are involved in important biochemical processes and related to various diseases. They have unrelated sequences, various substrates, and different kinds of catalytic activities, yet they share the same canonical alpha/beta hydrolase fold, which consists of an eight stranded parallel alpha/beta structure. They are also characterized by a catalytic triad composed of a histidine, an acid and a nucleophile. Members of this superfamily are often drug targets for treating diseases, such as diabetes, Alzheimer's disease, obesity and blood clotting disorders. The abhydrolase domain-containing protein 9 (ABHD9), also designated epoxide hydrolase 3 (EPHX3), is a 360 amino acid protein and a member of the alpha/beta hydrolase superfamily. The gene encoding ABHD9 maps to chromosome 19, which consists of over 63 million bases, houses approximately 1,400 genes and is recognized for having the greatest gene density of the human chromosomes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABHD9
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EPHX3
Gene Accession No. Q9H6B9
Gene Alias 2310063B19Rik; Abhd9; abhydrolase domain containing 9; abhydrolase domain-containing protein 9; EH3; Ephx3; Epoxide hydrolase 3
Gene Symbols EPHX3
Host Species Rabbit
Immunogen A synthesized peptide derived from human EPHX3(Accession Q9H6B9), corresponding to amino acid residues A37-L87.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79852
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.