missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABI1 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-59843G
This item is not returnable.
View return policy
Description
ABI1 Polyclonal antibody specifically detects ABI1 in Human samples. It is validated for Western Blot
Specifications
| ABI1 | |
| Polyclonal | |
| Western Blot | |
| Abelson interactor 1, ABI-1, abl interactor 1, Abl-binding protein 4, AblBP4, abl-interactor 1, Abl-interactor protein 1 long, E3B1, eps8 binding protein, Eps8 SH3 domain-binding protein, Eps8-binding protein, interactor protein AblBP4, nap1 binding protein, Nap1-binding protein, Nap1BP, spectrin SH3 domain binding protein 1, Spectrin SH3 domain-binding protein 1, SSH3BP, SSH3BP1 | |
| The immunogen recognized by this antibody maps to a region between residue 150 and 200 of human abl-interactor 1 using the numbering given in entry NP_005461.2 (GeneID 10006). | |
| 0.1 mL | |
| Phospho Specific | |
| 10006 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction