missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABI2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98107-100ul
This item is not returnable.
View return policy
Description
ABI2 Polyclonal antibody specifically detects ABI2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ABI2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Abelson interactor 2, Abi-2, ABI2B, abl binding protein 3, abl interactor 2, Abl-binding protein 3, AblBP3ABI-2, abl-interacting protein 1 (SH3-containing protein), abl-interactor 2, abl-interactor protein 2b, AIP-1, arg protein tyrosine kinase-binding protein, Arg-binding protein 1, argBP1, ARGBPIA, argBPIB, SSH3BP2 | |
| Produced in rabbits immunized with E. coli-derived Human ABI2 fragment. | |
| 100 μg | |
| Cancer | |
| 10152 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction