missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABI2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14890-1-AP-20UL
This item is not returnable.
View return policy
Description
Interaction of Fas ligand (TNFSF6) with the FAS antigen (TNFRSF6) mediates programmed cell death, also called apoptosis, in a number of organ systems. The protein encoded by this gene binds to FAS antigen and can initiate apoptosis or enhance apoptosis initiated through FAS antigen. Initiation of apoptosis by the protein encoded by this gene requires a ubiquitin-like domain but not the FAS-binding domain.Specifications
| ABI2 | |
| Polyclonal | |
| Unconjugated | |
| ABI2 | |
| Abelson interactor 2, ABI 2, ABI2, ABI2B, Abl binding protein 3, abl interactor 2, AblBP3, AIP 1, Arg binding protein 1, ArgBP1, argBPIA, argBPIB, SSH3BP2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10152, 286928, 329165 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P62484, Q9NYB9 | |
| Abi2 | |
| ABI2 Fusion Protein Ag6679 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction