missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABL2 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-18875DL594
This item is not returnable.
View return policy
Description
ABL2 Polyclonal antibody specifically detects ABL2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| ABL2 | |
| Polyclonal | |
| Western Blot | |
| Abelson murine leukemia viral (v-abl) oncogene homolog 2, Abelson murine leukemia viral oncogene homolog 2, Abelson-related gene protein, Abelson-relatedgene), arg tyrosine kinase, EC 2.7.10, EC 2.7.10.2, FLJ22224, FLJ31718, FLJ41441, tyrosine-protein kinase ABL2, Tyrosine-protein kinase ARG, v-abl Abelson murine leukemia viral oncogene homolog 2 | |
| The immunogen recognized by this antibody maps to a region between residue 750 and 800 of human Abelson-related gene protein using the numbering given in entry NP_009298.1 (GeneID 27). | |
| 0.1 mL | |
| Protein Kinase | |
| 27 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction