Learn More
Abnova™ CEN19q (DEAC) FISH Probe
Product Code. 16062735
Click to view available options
Quantity:
20 μL
Unit Size:
20µL
Product Code. 16062735
Brand: Abnova™ FC0159.20uL
Includes
FISH Hybridization Buffer (80μL)
Labeled FISH probes as chromosomal markers using Fluorescent In Situ Hybridization Technique.
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity
Specifications
Content And Storage Store at 4°C in the dark.
Label or Dye DEAC
For Use With (Application) Fluorescent In Situ Hybridization (Cell)
Includes FISH Hybridization Buffer (80 uL)
Location 19q12
Probe CEN19q
Product Type CEN19q (DEAC) FISH Probe
Quantity 20 μL