Learn More
Abnova™ Fluorescent Dye 700-M Succinimidyl Ester (Red)
Description
All the Fluorescent Dye M series products are designed to be maximally excited by one of the major light sources equipped in flow cytometers. Besides, in comparison with phycobiliprotein tandems, Fluorescent Dye M series possesses better photostability, higher conjugation yields, and little pH sensitivity making it an ideal choice in fluorescence imaging applications. Ideal for labeling of biological molecules for fluorescence imaging and other fluorescence-based biochemical analysis.
- Purity: >90%
- Spectral Equivalent: APC-Alexa Fluor 700 Tandems
- Molar Extinction Co-efficient: >2,50,000 M-1cm-1
- Excitation (Max): 657nm
- Emission (Max): 700nm
- Recommended Excitation: Red (633nm)
- Recommended Filter Sets: Cy5.5 Filter
- Solubility: DMSO, water
- Storage Buffer: DMSO
Application: Conjugation, Flow Cytometry
Recommended Storage: Store in desiccated environment at -20°C and avoid from light.
Specifications
Specifications
| Description | Fluorescent dye that is water-soluble and forms stable peptide bond with protein amino groups. All the Fluorescent Dye M series products are designed to be maximally excited by one of the major light sources equipped in flow cytometers. Besides, in comparison with phycobiliprotein tandems, Fluorescent Dye M series possesses better photostability, higher conjugation yields, and little pH sensitivity making it an ideal choice in fluorescence imaging applications. |
| For Use With (Application) | Conjugation; Flow Cytometry |
| Quantity | 1 mg |
| Product Type | Fluorescent Dye 700-M Succinimidyl Ester (Red) |
| Filter Type | Cy5.5 Filter |
| Purity | >90% |
| Equivalence | APC-Alexa Fluor 700 Tandems |
| Storage Buffer | DMSO |
| Content And Storage | Store the product in desiccated environment at -20°C and avoid from light. |
| Label or Dye | Fluorescent Dye 700-M Succinimidyl Ester (Red) |
| Show More |
For research use only (RUO).
Your input is important to us. Please complete this form to provide feedback related to the content on this product.