missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABO, Blood Group A Antigen Antibody (3-3A), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47872AF405
This item is not returnable.
View return policy
Description
ABO, Blood Group A Antigen Monoclonal antibody specifically detects ABO, Blood Group A Antigen in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ABO, Blood Group A Antigen | |
| Monoclonal | |
| Alexa Fluor 405 | |
| A transferase, A3GALNT, A3GALT1, ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase, transferase B, alpha 1-3-galactosyltransferase), ABO glycosyltransferase, B transferase, B(A) alpha-1,3-galactosyltransferase, EC 2.4.1.37, EC 2.4.1.40, Fucosylglycoprotein 3-alpha-galactosyltransferase, Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase, Glycoprotein-fucosylgalactoside alpha-galactosyltransferase, Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase, GTB, Histo-blood group A transferase, histo-blood group A2 transferase, histo-blood group ABO system transferase, Histo-blood group B transferase, NAGAT | |
| Mucin isolated from an ovarian cyst fluid (Uniprot: P16442) | |
| 0.1 mL | |
| Immunology | |
| 28 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 3-3A | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction