missing translation for 'onlineSavingsMsg'
Learn More

ABO, Blood Group A Antigen Antibody (VA4-20B) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

€ 201.00 - € 356.00

Specifications

Antigen ABO, Blood Group A Antigen
Clone VA4-20B
Concentration 1 mg/mL
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500
Applications Western Blot, Immunohistochemistry, Immunofluorescence
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18656726
View Documents
Novus Biologicals
NBP2-50485-0.025ml
0.025 mL
€ 201.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18670357
View Documents
Novus Biologicals
NBP2-50485
0.1 mL
€ 356.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ABO, Blood Group A Antigen Monoclonal antibody specifically detects ABO, Blood Group A Antigen in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

ABO, Blood Group A Antigen
1 mg/mL
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
Mouse
Immunology
PBS
28
IgG1
Protein A purified
VA4-20B
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500
Monoclonal
Purified
RUO
Human
A transferase, A3GALNT, A3GALT1, ABO blood group (transferase A, alpha 1-3-N-acetylgalactosaminyltransferase, transferase B, alpha 1-3-galactosyltransferase), ABO glycosyltransferase, B transferase, B(A) alpha-1,3-galactosyltransferase, EC 2.4.1.37, EC 2.4.1.40, Fucosylglycoprotein 3-alpha-galactosyltransferase, Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase, Glycoprotein-fucosylgalactoside alpha-galactosyltransferase, Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase, GTB, Histo-blood group A transferase, histo-blood group A2 transferase, histo-blood group ABO system transferase, Histo-blood group B transferase, NAGAT
Partially purified ABO, Blood Group A Antigen
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.