missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABRO Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-60449AF700
This item is not returnable.
View return policy
Description
ABRO Polyclonal antibody specifically detects ABRO in Human samples. It is validated for Western Blot
Specifications
| ABRO | |
| Polyclonal | |
| Western Blot | |
| Abraxas brother protein 1, ABRO1abraxas brother 1, Em:AC068896.4, family with sequence similarity 175, member B, FLJ22338, KIAA0157BRISC complex subunit Abro1, Protein FAM175B | |
| The immunogen recognized by this antibody maps to a region between residue 390 and 415 of human abraxas brother 1 using the numbering given in entry NP_115558.3 (GeneID 23172). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 23172 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction