missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABRO Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-60449
This item is not returnable.
View return policy
Description
ABRO Polyclonal specifically detects ABRO in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| ABRO | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| Abraxas brother protein 1, ABRO1abraxas brother 1, Em:AC068896.4, family with sequence similarity 175, member B, FLJ22338, KIAA0157BRISC complex subunit Abro1, Protein FAM175B | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 23172 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
| NP_115558.3 | |
| ABRAXAS2 | |
| The immunogen recognized by this antibody maps to a region between residue 390 and 415 of human abraxas brother 1 using the numbering given in entry NP_115558.3 (GeneID 23172). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction