Learn More
ABT-046, MedChemExpress
Product Code. 16406368
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16406368
Brand: Medchem Express HY15197
MedChemExpress ABT-046 is a potent, selective, and orally efficacious acyl CoA:diacylglycerol acyltransferase 1 (DGAT-1) inhibitor (IC50= 8 nM).
ABT-046 is a potent, selective, and orally efficacious acyl CoA:diacylglycerol acyltransferase 1 (DGAT-1) inhibitor (IC50= 8 nM).
Specifications
Chemical Name or Material ABT-046
CAS 1031336-60-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H22N4O2
Quantity 10 mg
Solubility Information DMSO : 66.67 mg/mL (190.26 mM; Need ultrasonic)
SMILES NC1=C(C2=CC=C([C@H]3CC[C@H](CC(O)=O)CC3)C=C2)C=NC4=CC=NN41
Molecular Weight (g/mol) 350.41
Formula Weight 350.41
Percent Purity 98.13%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only