missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ABT1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22332UV
This item is not returnable.
View return policy
Description
ABT1 Polyclonal antibody specifically detects ABT1 in Human samples. It is validated for Western Blot
Specifications
| ABT1 | |
| Polyclonal | |
| Western Blot | |
| activator of basal transcription 1, Basal transcriptional activator, hABT1, TATA-binding protein-binding protein | |
| The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Activator of Basal Transcription 1 using the numbering given in entry NP_037507.1 (GeneID 29777). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 29777 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction