missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ABT1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16361965
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16361965 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16361965 Supplier Invitrogen™ Supplier No. PA585017

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SKKRVVPGIV YLGHIPPRFR PLHVRNLLSA YGEVGRVFFQ AEDRFVRRKK KAAAAAGGKK RSYTKDYTEG WVEFR Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 89%.

Could be a novel TATA-binding protein (TBP) which can function as a basal transcription activator. Can act as a regulator of basal transcription for class II genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ABT1
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Abt1
Gene Accession No. Q9ULW3
Gene Alias ABT1; Activator of basal transcription 1; basal transcriptional activator; hABT1; hypothetical protein LOC553568; im:7139827; TATA-binding protein-binding protein; zgc:109995
Gene Symbols Abt1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ABT1. Recombinant protein control fragment (Product #RP-108674).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29777
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.