missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ABTB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565075
This item is not returnable.
View return policy
Description
Immunogen sequence: MVDTRISVRIH EYAAISLTAC MENLVEEIRA RVMASHSPDG GGAGGGEVSA EALEMVINND AELWGVLQPY EH Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.
May be involved in the initiation of hepatocyte growth. Target protein displays a transient increase in mRNA expression prior to DNA synthesis in regenerating liver cells.
Specifications
| ABTB2 | |
| Polyclonal | |
| Unconjugated | |
| ABTB2 | |
| ABTB2; ABTB2A; ankyrin repeat and BTB (POZ) domain containing 2; ankyrin repeat and BTB domain containing 2; ankyrin repeat and BTB/POZ domain-containing protein 2; BTBD22 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25841 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N961 | |
| ABTB2 | |
| Recombinant Human ABTB2. Recombinant protein control fragment (Product #RP-105837). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction