Learn More
Tocris Bioscience™ AC 265347
GREENER_CHOICE
CaSR biased allosteric modulator
Brand:  Tocris Bioscience™ 6165/50
Encompass
Product Code. 15766369
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
AC 265347 is a CaSR biased allosteric modulator (pEC50 = 7.8-8.1). Calcimimetic. Displays ability to bias signalling towards the accumulation of pERK1/2 and IP1. Reduces parathyroid hormone (PTH) levels in rat serum without inducing the release of calcitonin. Exhibits no significant activation of human GABAB or type I PTH receptors. Orally active.
Specifications
Specifications
α-(2,4-Dimethylphenyl)-α-methyl-2-benzothiazolemethanol
1253901-26-6
>99%