Learn More
Ac-Asp-Glu-Val-Asp-7-Amino-4-TriFluoromethylcoumarin, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
- Ac-DEVD-AFC is a sensitive fluorescent substrate for the assay of caspase-3 and CPP-32 activity.
- The trifluoromethyl substituent of the fluorophore improves the membrane permeability of the substrate.
- Protein or Enzyme Type: Activators, Inhibitors and Substrates
Specifications
Specifications
| Form | Lyophilized |
| Quantity | 10 mg |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.