Learn More
Tocris Bioscience™ Ac-Cys-NHMe
GREENER_CHOICE
Product Code. 30000012
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30000012 10 mg 10mg
30000025 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30000012 Supplier Tocris Bioscience™ Supplier No. 7936/10
  • ONLINE EXCLUSIVE PRICE
    € 161.00
    € 152.25 / 10mg
    Save € 8.75
    5% Off
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
Building block for the lysine acylation using conjugating enzymes (LACE) technique
Ac-Cys-NHMe is a non-canonical amino acid that is an essential building block for lysine acylation using conjugating enzymes (LACE) technique. LACE is a chemoenzymatic approach that uses an E2 small ubiquitin-like modifier (SUMO)-conjugating enzyme, Ubc9, to conjugate peptide thioesters containing a C-terminal ubiquitin-derived sequence (LRLRGG) to a lysine residue located in a recognition sequence (IKQE) on a protein via an isopeptide bond. Ac-Cys-NHMe can be reacted with a peptide hydrazide to form a reactive peptide thioester, which is the substrate for Ubc9. LACE enables the site-specific modification of proteins at lysine residues to allow attachment of biochemical probes such as fluorophores to proteins or to engineer proteins for site-specific delivery.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2R)-2-(Acetylamino)-3-mercapto-N-methylpropanamide
CAS 10061-65-1
Quantity 10 mg
Target Unnatural Amino Acids
Molecular Formula C6H12N2O2S