missing translation for 'onlineSavingsMsg'
Learn More

Ac-Glu-OMe, TRC

Product Code. 30321486
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30321486 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321486 Supplier Toronto Research Chemicals Supplier No. TRCA16920510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ac-Glu-OMe
CAS 17015-15-5
Molecular Formula C8H13NO5
Quantity 10 mg
SMILES COC(=O)[C@H](CCC(=O)O)NC(=O)C
Molecular Weight (g/mol) 203.19
Formula Weight 203.0794
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.