Learn More
ACAA2 Monoclonal Antibody (OTI1B1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA506129S
Encompass_Preferred
 View more versions of this product
Product Code. 17525502
  • € 242.00 / 30µL
Description
ACAA2 Monoclonal antibody specifically detects ACAA2 in Canine, Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Specifications
Specifications
ACAA2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P13437, P42765, Q8BWT1
Mouse
Affinity chromatography
RUO
10449, 170465, 490568, 52538
-20° C, Avoid Freeze/Thaw Cycles
IgG2b
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
OTI1B1
Unconjugated
ACAA2
0610011L04Rik; 3-ketoacyl-CoA thiolase, mitochondrial; Acaa2; Acetyl-CoA acetyltransferase; Acetyl-CoA acyltransferase; acetyl-CoA acyltransferase 2; acetyl-Coenzyme A acyltransferase 2; acetyl-Coenzyme A acyltransferase 2 (mitochondrial 3-oxoacyl-Coenzyme A thiolase); Acyl-CoA hydrolase, mitochondrial; AI255831; AI265397; beta ketothiolase; beta-ketothiolase; D18Ertd240e; DSAEC; mitochondrial 3-oxoacyl-CoA thiolase; mitochondrial 3-oxoacyl-Coenzyme A thiolase; T1
Full length human recombinant protein of human ACAA2 produced in HEK293T cell.
30 μL
Primary
Canine, Human, Mouse, Rat