Learn More
Invitrogen™ ACAA2 Recombinant Rabbit Monoclonal Antibody (23GB2955), Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553590
 View more versions of this product
Product Code. 30089872
  • € 562.00 / 100µL
Description
ACAA2 (3-Ketoacyl-CoA thiolase) is an enzyme that in humans is encoded by the ACAA2 gene. Acetyl-Coenzyme A acyltransferase 2 is an acetyl-CoA C-acyltransferase enzyme. The encoded protein catalyzes the last step of the mitochondrial fatty acid beta-oxidation spiral. Unlike most mitochondrial matrix proteins, it contains a non-cleavable amino-terminal targeting signal.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ACAA2
Recombinant Monoclonal
Unconjugated
ACAA2
0610011L04Rik; 3-ketoacyl-CoA thiolase, mitochondrial; Acaa2; Acetyl-CoA acetyltransferase; Acetyl-CoA acyltransferase; acetyl-CoA acyltransferase 2; acetyl-Coenzyme A acyltransferase 2; acetyl-Coenzyme A acyltransferase 2 (mitochondrial 3-oxoacyl-Coenzyme A thiolase); Acyl-CoA hydrolase, mitochondrial; AI255831; AI265397; beta ketothiolase; beta-ketothiolase; D18Ertd240e; DSAEC; mitochondrial 3-oxoacyl-CoA thiolase; mitochondrial 3-oxoacyl-Coenzyme A thiolase; T1
Rabbit
Affinity chromatography
RUO
10449, 170465, 52538
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
23GB2955
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P13437, P42765, Q8BWT1
ACAA2
A synthesized peptide derived from ACAA2 (360-397AA).