missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACAA2 Recombinant Rabbit Monoclonal Antibody (8H6H5)

Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA542617
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Positive test controls include: HeLa, HepG2, A-549, Rat heart. The target is usually found in the following locations: Mitochondrion. Immunogen sequence: PAISGALKKA GLSLKDMDLV EVNEAFAPQY LAVERSLDLD ISKTNVNGGA IALGHPLGGS GSRITAHLVH ELRRRGGKYA VGSACIGGGQ GIAVIIQSTA.
ACAA2 (3-Ketoacyl-CoA thiolase) is an enzyme that in humans is encoded by the ACAA2 gene. Acetyl-Coenzyme A acyltransferase 2 is an acetyl-CoA C-acyltransferase enzyme. The encoded protein catalyzes the last step of the mitochondrial fatty acid beta-oxidation spiral. Unlike most mitochondrial matrix proteins, it contains a non-cleavable amino-terminal targeting signal.
Spécification
| ACAA2 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P13437, P42765, Q8BWT1 | |
| ACAA2 | |
| A synthetic peptide corresponding to a sequence within amino acids 298-397 of human ACAA2 (P42765). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| 8H6H5 | |
| Unconjugated | |
| ACAA2 | |
| 0610011L04Rik; 3-ketoacyl-CoA thiolase, mitochondrial; Acaa2; Acetyl-CoA acetyltransferase; Acetyl-CoA acyltransferase; acetyl-CoA acyltransferase 2; acetyl-Coenzyme A acyltransferase 2; acetyl-Coenzyme A acyltransferase 2 (mitochondrial 3-oxoacyl-Coenzyme A thiolase); Acyl-CoA hydrolase, mitochondrial; AI255831; AI265397; beta ketothiolase; beta-ketothiolase; D18Ertd240e; DSAEC; mitochondrial 3-oxoacyl-CoA thiolase; mitochondrial 3-oxoacyl-Coenzyme A thiolase; T1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10449, 170465, 52538 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu