missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACACB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552309
This item is not returnable.
View return policy
Description
Immunogen sequence: ITKSKSEANL IPSQEPFPAS DNSGETPQRN GEGHTLPKTP SQAEPASHKG PKDAGRRRNS LPPSHQKPPR NPLSSSDAAP SPELQANGTG TQGLEATDTN GLSSSARPQG QQAGSPSKED KKQANIKRQL MTNFILGSFD DYSS Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 57%.
Catalyzes the ATP-dependent carboxylation of acetyl-CoA to malonyl-CoA. Carries out three functions: biotin carboxyl carrier protein, biotin carboxylase and carboxyltransferase. Involved in inhibition of fatty acid and glucose oxidation and enhancement of fat storage. May play a role in regulation of mitochondrial fatty acid oxidation through malonyl-CoA-dependent inhibition of carnitine palmitoyltransferase 1.
Specifications
| ACACB | |
| Polyclonal | |
| Unconjugated | |
| ACACB | |
| Acacb; Acc2; ACCB; ACC-beta; acetyl-CoA carboxylase 2; acetyl-CoA carboxylase beta; acetyl-Coenzyme A carboxylase 2; acetyl-Coenzyme A carboxylase beta; AI597064; AW743042; Biotin carboxylase; HACC275 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 32 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O00763 | |
| ACACB | |
| Recombinant protein corresponding to Human ACACB. Recombinant protein control fragment (Product #RP-102319). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction