missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACADL Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97409APC
This item is not returnable.
View return policy
Description
ACADL Polyclonal antibody specifically detects ACADL in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ACADL | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| ACAD4, acyl-CoA dehydrogenase, long chain, acyl-Coenzyme A dehydrogenase, long chain, EC 1.3.99, EC 1.3.99.13, LCADFLJ94052, long-chain specific acyl-CoA dehydrogenase, mitochondrial | |
| Produced in rabbits immunized with E. coli-derived Human ACADL fragment. | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 33 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction