missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACADS Antibody (JE64-13), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-31991
This item is not returnable.
View return policy
Description
ACADS Monoclonal antibody specifically detects ACADS in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| ACADS | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:100 | |
| ACAD3, acyl-CoA dehydrogenase, C-2 to C-3 short chain, acyl-Coenzyme A dehydrogenase, C-2 to C-3 short chain, Butyryl-CoA dehydrogenase, EC 1.3.99, EC 1.3.99.2, mitochondrial short-chain specific acyl-CoA dehydrogenase, SCADshort-chain acyl-CoA dehydrogenase, short-chain specific acyl-CoA dehydrogenase, mitochondrial, unsaturated acyl-CoA reductase | |
| Synthetic peptide within human ACADS aa 363-412/412. (Uniprot: P16219) | |
| 100 μL | |
| Signal Transduction | |
| 35 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| JE64-13 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction