missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACADS Mouse anti-Human, Clone: OTI1D2, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800429S
This item is not returnable.
View return policy
Description
ACADS Monoclonal antibody specifically detects ACADS in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| ACADS | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P16219 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 35 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1D2 | |
| Unconjugated | |
| ACADS | |
| ACAD3; Acads; acetyl-Coenzyme A dehydrogenase, short chain; acyl-CoA dehydrogenase short chain; acyl-CoA dehydrogenase, C-2 to C-3 short chain; acyl-Coenzyme A dehydrogenase, C-2 to C-3 short chain; acyl-Coenzyme A dehydrogenase, short chain; AI196007; Bcd1; Bcd-1; butyryl-CoA dehydrogenase; EGK_04272; Hdlq8; mitochondrial short-chain specific acyl-CoA dehydrogenase; SCAD; short chain acyl-coenzyme A dehydrogenase; short-chain acyl-CoA dehydrogenase; short-chain specific acyl-CoA dehydrogenase, mitochondrial; unsaturated acyl-CoA reductase | |
| Human recombinant protein fragment corresponding to amino acids 34-260 of human ACADS produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction