missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ACADVL Recombinant Rabbit Monoclonal Antibody (23GB3615), Invitrogen™
GREENER_CHOICE

Product Code. 30087688
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30087688 100 μL 100µL
30089957 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30087688 Supplier Invitrogen™ Supplier No. MA553458

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

The protein encoded by this gene is targeted to the inner mitochondrial membrane where it catalyzes the first step of the mitochondrial fatty acid beta-oxidation pathway. This acyl-Coenzyme A dehydrogenase is specific to long-chain and very-long-chain fatty acids. A deficiency in this gene product reduces myocardial fatty acid beta-oxidation and is associated with cardiomyopathy. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACADVL
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3615
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ACADVL
Gene Accession No. P49748
Gene Alias ACAD6; Acadvl; acyl-CoA dehydrogenase, very long chain; acyl-Coenzyme A dehydrogenase, very long chain; LCACD; MVLCAD; Very long chain Acyl-Coa dehydrogenase; very long-chain specific acyl-CoA dehydrogenase, mitochondrial; Vlcad; VLCAD very-long-chain acyl-CoA dehydrogenase
Gene Symbols ACADVL
Host Species Rabbit
Immunogen A synthetic peptide of human ACADVL/VLCAD (25-79AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 37
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.