missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ACAP2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16536631
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16536631 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16536631 Supplier Invitrogen™ Supplier No. PA588364

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: KAFCVANKQF MNGIRDLAQY SSNDAVVETS LTKFSDSLQE MINFHTILFD QTQRSIKAQL QNFVKEDLRK FKDAKKQFEK VSEEKENALV KNAQVQRNKQ HEVEEATNIL T; Positive Samples: LO2, HeLa, Jurkat, U-87MG, A-549, Mouse brain, Mouse liver; Cellular Location: Endosome membrane, Peripheral membrane protein.

ACAP2 is a transport carrier belonging to the Centaurin family with three ANK repeats, an Arf-GAP domain, a BAR domain, and a PH domain. ACAP2, a GTPase-activating protein (GAP) for ADP ribosylation factor 6 (ARF6) has robust constitutive ARF6 GAP activity, where the GAP activity is stimulated by phosphatidylinositol 4,5-bisphosphate (PIP2) and phosphatidic acid but has little activity toward ARF1 and is known to control the return of ARF to the inactive GDB-bound shots. ACAP2, along with ACAP1, is targeted by phosphatidyl inositol (3, 4, 3) triphosphate inhibiting the formation of PDGF-induced dorsal membrane ruffles in NIH3T3 fibroblasts when overexpressed. ACAP2 is phosphorylated upon DNA damage, probably by ATM or ATR. Expression is wide-spread with the highest level in lung.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACAP2
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ACAP2
Gene Accession No. Q15057, Q6ZQK5
Gene Alias 4832442G16; 9530039J15Rik; ACAP2; Arf GAP with coiled coil, ANK repeat and PH domains 2; arf-GAP with coiled-coil, ANK repeat and PH domain-containing protein 2; ArfGAP with coiled-coil, ankyrin repeat and PH domains 2; centaurin beta2; centaurin, beta 2; centaurin-beta-2; Centb2; CNT-B2; KIAA0041; mKIAA0041
Gene Symbols ACAP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 50-160 of human ACAP2 (NP_0364193).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23527, 78618
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.