missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACAT1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-46140
This item is not returnable.
View return policy
Description
ACAT1 Polyclonal specifically detects ACAT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Peptide ELISA.
Specifications
| ACAT1 | |
| Polyclonal | |
| Western Blot 0.01 - 0.03 ug/ml, Immunocytochemistry/Immunofluorescence, Peptide ELISA Detection limit 1:2000 | |
| P24752 | |
| ACAT1 | |
| Peptide with sequence C-DEEYKRVDFSKVPK corresponding to internal region according to NP_000010.1. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody recognizes and epitope within the internal region. Cross reacts with Human, Mouse and Rat. Expected from sequence similarity: Dog and Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence, ELISA | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| ACAT, acetoacetyl Coenzyme A thiolase, Acetoacetyl-CoA thiolase, acetyl-CoA acetyltransferase 1, acetyl-CoA acetyltransferase, mitochondrial, acetyl-Coenzyme A acetyltransferase 1, EC 2.3.1, EC 2.3.1.9, MAT, mitochondrial acetoacetyl-CoA thiolase, T2, THIL | |
| Goat | |
| 45 kDa | |
| 0.1 mg | |
| Cancer, Cellular Markers, Lipid and Metabolism, Mitochondrial Markers, Signal Transduction | |
| 38 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction