missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACAT2 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32198C
This item is not returnable.
View return policy
Description
ACAT2 Polyclonal antibody specifically detects ACAT2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ACAT2 | |
| DyLight 650 | |
| Rabbit | |
| Cholesterol Metabolism, Lipid and Metabolism | |
| Store at 4°C in the dark. |
| Western Blot | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| 0.1 mL | |
| Mouse |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction