missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACAT2 Monoclonal Antibody (OTI5F7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501219
This item is not returnable.
View return policy
Description
The product of this gene is an enzyme involved in lipid metabolism, and it encodes cytosolic acetoacetyl-CoA thiolase. This gene shows complementary overlapping with the 3-prime region of the TCP1 gene in both mouse and human. These genes are encoded on opposite strands of DNA, as well as in opposite transcriptional orientation.Specifications
| ACAT2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q5XI22, Q9BWD1 | |
| Acat2 | |
| Full length human recombit protein of human ACAT2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Rat | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI5F7 | |
| Unconjugated | |
| ACAT2 | |
| acetyl-CoA acetyltransferase 2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 308100, 39, 484063 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction