missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACBD3 Antibody (OTI1G2), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72148DL594
This item is not returnable.
View return policy
Description
ACBD3 Monoclonal antibody specifically detects ACBD3 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry, Immunohistochemistry
Specifications
| ACBD3 | |
| Monoclonal | |
| DyLight 594 | |
| acyl-CoA binding domain containing 3, Acyl-CoA-binding domain-containing protein 3, acyl-Coenzyme A binding domain containing 3, GCP60Golgi phosphoprotein 1, GOCAP1Peripheral benzodiazepine receptor-associated protein PAP7, golgi complex associated protein 1, 60kDa, Golgi complex-associated protein 1, Golgi resident protein GCP60, GOLPH1PBR- and PKA-associated protein 7, PAP7, PBR associated protein, peripherial benzodiazepine receptor associated protein, PKA (RIalpha)-associated protein | |
| Full length recombinant protein of human ACBD3 (NP_073572) produced in HEK293T cells. | |
| 0.1 mL | |
| Golgi Apparatus Markers | |
| 64746 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry | |
| OTI1G2 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction