missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACBD3, Mouse anti-Human, Clone: 1E10, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00064746-M05.50ug
This item is not returnable.
View return policy
Description
Sequence: RRLEQRWGFGLEELYGLALRFFKEKDGKAFHPTYEEKLKLVALHKQVLMGPYNPDTCPEVGFFDVLGNDRRREWAALGNMSKEDAMVEFVKLLNRCCHLSpecifications
| ACBD3 | |
| Monoclonal | |
| Unconjugated | |
| acyl-Coenzyme A binding domain containing 3 | |
| GCP60/GOCAP1/GOLPH1/PAP7 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1E10 | |
| In 1x PBS, pH 7.4 | |
| NM_022735 | |
| ACBD3 | |
| ACBD3 (NP_073572, 73 a.a. ∼ 171 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 64746 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction