missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACBD5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97727-100ul
This item is not returnable.
View return policy
Description
ACBD5 Polyclonal antibody specifically detects ACBD5 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ACBD5 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| acyl-CoA binding domain containing 5, acyl-Coenzyme A binding domain containing 5, DKFZp434A2417, endozepine-related protein, KIAA1996acyl-CoA-binding domain-containing protein 5, membrane-associated diazepam binding inhibitor | |
| Produced in rabbits immunized with E. coli-derived Human ACBD5 fragment. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 91452 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction