missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACBD5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
€ 292.00 - € 549.00
Specifications
| Antigen | ACBD5 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18436621
|
Novus Biologicals
NBP1-85146-25ul |
25ul |
€ 292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18738373
|
Novus Biologicals
NBP1-85146 |
€ 549.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
ACBD5 Polyclonal specifically detects ACBD5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ACBD5 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 91452 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:IQVPPGNGNIGNMQVVAVEGKGEVKHGGEDGRNNSGAPHREKRGGETDEFSNVRRGRGHRMQHLSEGTKGRQVGSGGDGERWGSDRGSRGSLNEQIALVLMRLQEDMQNVLQRLQKLETLTALQAKSSTSTLQTAPQPTSQRPSWWPF | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| acyl-CoA binding domain containing 5, acyl-Coenzyme A binding domain containing 5, DKFZp434A2417, endozepine-related protein, KIAA1996acyl-CoA-binding domain-containing protein 5, membrane-associated diazepam binding inhibitor | |
| ACBD5 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title