missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Antibody detects endogenous levels of total ACBD5.
Acyl-CoA binding protein which acts as the peroxisome receptor for pexophagy but is dispensable for aggrephagy and nonselective autophagy. Binds medium- and long-chain acyl-CoA esters.
Specifications
Specifications
| Antigen | ACBD5 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | ACBD5 |
| Gene Accession No. | A0FKI7, Q5T8D3, Q5XG73 |
| Gene Alias | 1300014E15Rik; Acbd5; acyl-CoA binding domain containing 5; acyl-CoA-binding domain-containing protein 5; acyl-Coenzyme A binding domain containing 5; endozepine-related protein; KIAA1996; membrane-associated diazepam binding inhibitor |
| Gene Symbols | ACBD5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
