missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ACBD6 Antibody Pair [HRP]

Product Code. 18209267 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18209267 15 Plates Pack of 15
18615781 5 Plates Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18209267 Supplier Novus Biologicals™ Supplier No. NBP27934715PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human ACBD6.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 46.875 pg/mL
Assay Sensitivity 46.88-3000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human ACBD6.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias acyl-CoA binding domain containing 6, acyl-CoA-binding domain-containing protein 6, acyl-Coenzyme A binding domain containing 6, MGC2404
Gene ID (Entrez) 84320
Gene Symbol ACBD6
Host Species Mouse
Kit Contents
  • Detection Antibody: 0.5 mg/mL mouse anti-ACBD6 monoclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Capture Antibody: 0.4 mg/mL of mouse anti-ACBD6 monoclonal antibody.
  • Standard: Each vial contains 150 ng of recombinant ACBD6.
  • Quantity 15 Plates
    Regulatory Status RUO
    Storage Requirements Storage is content dependent.
    Target ACBD6
    Technique 1. Capture Antibody: Dilute to a working concentration of 2.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 3000 pg/mL is recommended.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.