Learn More
ACBP Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-97012UV
Product Code. 18181250
  • € 415.00 / 0.10mL
Description
ACBP Polyclonal specifically detects ACBP in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ACBP
Polyclonal
50 mM Sodium Borate
Rabbit
Affinity purified
RUO
Primary
Human
Purified
ELISA
DyLight 350
acyl-Coenzyme A binding domain containing 1, acyl-Coenzyme A bindingprotein), CCK-RP, cholecystokinin-releasing peptide, trypsin-sensitive, diazepam binding inhibitor (GABA receptor modulator, acyl-CoA binding protein), Diazepam-binding inhibitor, endozepine, EP, GABA receptor modulator, MGC70414
Produced in rabbits immunized with purified, recombinant ACBP (P07108-2; Met1-Ala87).
0.1 mL
Cancer
1622
Store at 4°C in the dark.