missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACBP Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97012PCP
This item is not returnable.
View return policy
Description
ACBP Polyclonal antibody specifically detects ACBP in Human samples. It is validated for ELISA
Specifications
| ACBP | |
| Polyclonal | |
| ELISA | |
| acyl-Coenzyme A binding domain containing 1, acyl-Coenzyme A bindingprotein), CCK-RP, cholecystokinin-releasing peptide, trypsin-sensitive, diazepam binding inhibitor (GABA receptor modulator, acyl-CoA binding protein), Diazepam-binding inhibitor, endozepine, EP, GABA receptor modulator, MGC70414 | |
| Produced in rabbits immunized with purified, recombinant Human ACBP (Uniprot#: P07108-2; Met1-Ala87). | |
| 0.1 mL | |
| Cancer | |
| 1622 | |
| Store at 4C in the dark. | |
| IgG |
| ELISA | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu