Learn More
Thermo Scientific™ Access Key Options for Ultra-Low Temperature Freezers
Product Code. 13540770 Shop All Thermo Scientific Products
Click to view available options
For Use With (Equipment):
Revco™ UxF
TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE Series, Revco TLE Series
TSX Series, Revco RLE, HERAfreeze HLE, Forma 89000, TLE Series
TSX Universal
TSX Universal (-ULTS Models)
Thermo Scientific TSU Series
Type:
Access Key Pack (EU) Standalone Cards
Access Key Pack (U.S.) Standalone Cards
Factory installed. No stand-alone orders.
Factory-installed. No stand-alone orders.
Field installed
Field-installed
Unit Size:
Each
Pack of 5
Product Code. 13540770
Brand: Thermo Scientific™ FRAC34567
Only null left
Add to basket
Add to basket
Record who opened the ultra low freezer door, when it was opened and how long it was open with access key options.
Analyze records of which staff opened the freezer door, what time is was opened and for what duration with the Thermo Scientific™ Access Key Options for Ultra-Low Temperature Freezers.
Features
  • Card access comes with 5 key cards
  • Field installed - purchased separately
  • Factory installed in the plant with purchase of unit
TRUSTED_SUSTAINABILITY
Specifications
Type Field installed
For Use With (Equipment) Revco™ UxF
Includes Access Key Option; Card access control w/five key cards