Learn More
Merck Accessories for Analytical Filter Holders
Accessories for filter holders for sample preparation.
Brand: Merck XX5002501
146.51 EUR valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.10000kg
Description
Accessories for filter holders for sample preparation.
Specifications
Not Autoclavable | |
25 mm glass microanalysis vacuum filter | |
Non-sterile | |
25 mm |
Support Screen (for XX10 025 30) | |
Stainless Steel | |
Support Screen |