Learn More
Thermo Scientific™ Accessories for MaxQ™ 7000 Water Bath Shakers
Product Code. 10071612 Shop All Thermo Scientific Products
Click to view available options
Product Type:
Auxillary Platform Suspension Arms
Cooling Coil
Gable Cover
Movable Dolly
Plexiglas™ Gable Cover
Unit Size:
Each
Product Code. 10071612
Brand: Thermo Scientific™ SHK700032
Only null left
Add to basket
Add to basket
Optimize the Accessories for Thermo Scientific™ MaxQ™ 7000 Water Bath Shakers to increase the versatility of the MaxQ 7000.
Put Thermo Scientific™ Accessories for MaxQ™ 7000 Water Bath Shakers to use to increase the utility of MaxQ 7000 Water Bath Shakers. With exceptional innovation, reliability and accuracy, Thermo Scientific MaxQ 7000 Shakers, Platforms and accessories are the optimal choice for your busy lab.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Plexiglas™ Gable Cover
For Use With (Application) for temperatures up to 65°C
For Use With (Equipment) MaxQ 7000