Learn More
Thermo Scientific™ Accessories for MaxQ™ 7000 Water Bath Shakers
Product Code. 10245902 Shop All Thermo Scientific Products
Click to view available options
Product Type:
Auxillary Platform Suspension Arms
Cooling Coil
Gable Cover
Movable Dolly
Plexiglas™ Gable Cover
Unit Size:
Each
Product Code. 10245902
Brand: Thermo Scientific™ SHK700037
Optimize the Accessories for Thermo Scientific™ MaxQ™ 7000 Water Bath Shakers to increase the versatility of the MaxQ 7000.
Put Thermo Scientific™ Accessories for MaxQ™ 7000 Water Bath Shakers to use to increase the utility of MaxQ 7000 Water Bath Shakers. With exceptional innovation, reliability and accuracy, Thermo Scientific MaxQ 7000 Shakers, Platforms and accessories are the optimal choice for your busy lab.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cooling Coil
For Use With (Application) for near-ambient temperature and below (requires a circulating chiller)
For Use With (Equipment) MaxQ 7000