Learn More
Cytiva Accessories and Replacement Parts for Amersham™ VacuGene XL Vacuum Blotting System
Shop All Cytiva ProductsDescription
- Rapidly transfer DNA from agarose gels up to 20 x 30cm in less than 1 hour
- Efficient and reproducible transfers improve blotting results
- Carry out all pretransfer steps in the unit
- Precise vacuum control for optimal transfer conditions
- RNA transfers in less than 4 hours
Specifications
Specifications
| For Use With (Equipment) | Amersham VacuGene XL Vacuum Blotting System |
| Type | Gel Supporting Screen |