Learn More
WTW™ Accessories for WTW™ Portable pH Meter
Brand: WTW™ 103711
Additional Details : Weight : 0.10000kg
Specifications
S7 | |
PVC-U Armoring, Glass Shaft, PE Watering Cap, FPM O-ring | |
30°C | |
Measurements in waste water, emulsions, suspensions and media that contain protein and sulphide | |
-200 to +200 | |
30°C |
2 Holes | |
4 to 12 | |
ORP Combination Electrode | |
SensoLyt™ PtA | |
With watering cap, filled with KCl (3mol/L), Ag+ free |