Description
Suitable for all machines, width 90, depth 70 cm
Specifications
Specifications
| Material | Stainless Steel |
| Product Type | Plinth/Base |
| Color | Stainless Steel |
| For Use With (Equipment) | Large-capacity washer-disinfectors, laboratory |