missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PepMap™ 300 C18 HPLC Columns

Product Code. 11312213 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
5 mm
150 mm
Diameter (Metric):
0.075 mm
0.3 mm
1 mm
Column Format:
Analytical Column
Analytical Column, NanoViper
Trap Cartridge
Unit Size:
Each
Pack of 5
This item is not returnable. View return policy

Product Code. 11312213

Brand: Thermo Scientific™ 163574

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use this nanocolumn, packed for LC-MS/MS peptide mapping, protein identification, biomarker discovery, and systems biology applications.

This is the standard column for high-resolution, high-sensitivity protein identification for proteomics and biomarker discovery and verification. The Thermo Scientific™ Acclaim™ PepMap™ 300 C18 LC Columns is ideal for:

  • High-resolution peptide mapping of tryptic, natural, and synthetic peptides
  • LC-MS/MS peptide mapping for protein identification, biomarker discovery, and systems biology

High loading capacity

 

Acclaim PepMap columns are exceptionally suitable for the analysis of low abundant peptides often found in complex proteomics samples.

The Acclaim PepMap C18 300Å column 5μm Particle Size is available in 150mm lengths in 75, 300, 1000, and classic 75μm I.D. to allow for tuning throughput, sensitivity, and amount of sample available.

 

  • Superior protein and peptide separations
  • Trifluoroacetic acid (TFA)-free LC/MS minimizes suppression effects for enhanced MS sensitivity
  • Outstanding separation efficiency
  • Superior resolution and recovery
  • Easy installation with nanoViper™ Fingertight Fittings

Columns are available with Thermo Scientific™ Dionex™ nanoViper Fingertight Fittings:

  • Requires no tools
  • Provides virtually zero-dead-volume connections
  • Withstands pressures up to a 1000 bar
  • About Nano, capillary, and micro Acclaim PepMap columns

Nano (75μm), capillary (300μm) and micro (1000μm) columns are ideal for protein and peptide analyses. Sample amount is often limited and smaller I.D. columns can increase in sensitivity in sample-limited situations. While nano LC columns are often used in proteomics, the concentrating effects of capillary and micro columns are beneficial for metabolomics or for facilitating LC-MS interfacing. Thermo Scientific Acclaim PepMap columns are offered in nano, capillary, and micro column in a variety of chemistries for peptides and protein analysis.

TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 0.075 mm
Flow Rate 200 mL/min
Length (Metric) 150 mm
Max. Pressure 7250 psi (500 bar)
Particle Size 5 μm
pH 2 to 8
Pore Size 300 Å
Surface Area 100 m²/g
Temperature 60°C
USP Type L1
Stationary Phase C18
Product Line nanoLC
Packing Material Spherical, Fully Porous, Ultrapure Silica
Carbon Load 9%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Type Analytical Column
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.